L-Tyrosine methyl ester 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
H-Tyr-OMe
H62069-14
H62069-22
AAH62069-14EA
67.6
USD
AAH62069-14
AAH62069-22
L-Tyrosine methyl ester 98%
L-Tyrosine methyl ester
L-Tyrosine methyl ester, 98%
Purity: 98%
CAS No.: 1080-06-4
Molecular Formula: C10H13NO3
Formula Weight: 195.22
Storage Temperature: 2°C to 8°C
Melting Point: 134-136°
Beilstein Reference: 2372626
MDL No.: MFCD00002392
Formula:
C₁₀H₁₃NO₃ MW: 195.22 g/mol Melting Pt: 134…136 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002392 CAS Number: 1080-06-4 EINECS: 214-095-3 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...