tert-Butyl-3-oxocyclobutanecarboxylate 97%

Supplier: Thermo Scientific Chemicals

Synonyms: tert-Butyl 3-oxocyclobutanecarboxylate

H62064-03 H62064-06
AAH62064-03EA 204 USD
AAH62064-03 AAH62064-06 AAH62064-MD
tert-Butyl-3-oxocyclobutanecarboxylate 97%
tert-Butyl-3-oxocyclobutanecarboxylate

tert-Butyl 3-oxocyclobutanecarboxylate, 97%


Purity: 97% CAS No.: 145549-76-4 Molecular Formula: C9H14O3 Formula Weight: 170.20 MDL No.: MFCD12827541

Formula: C₉H₁₄O₃
MW: 170.208 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12827541
CAS Number: 145549-76-4

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR