trans-4-(Boc-amino)cyclohexanemethylamine 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: tert-Butyl (trans-4-(aminomethyl)cyclohexyl)carbamate

H62062-03 H62062-06
AAH62062-03EA 253 USD
AAH62062-03 AAH62062-06 AAH62062-MD
trans-4-(Boc-amino)cyclohexanemethylamine 97%
trans-4-(Boc-amino)cyclohexanemethylamine

trans-4-(Boc-amino)cyclohexanemethylamine, 97%


Purity: 97% CAS No.: 177583-27-6 Molecular Formula: C12H24N2O2 Formula Weight: 228.33 MDL No.: MFCD01861810

Formula: C₁₂H₂₄N₂O₂
MW: 228.336 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01861810
CAS Number: 177583-27-6

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR