Nω-Nitro-L-arginine 98%

Supplier: Thermo Scientific Chemicals

Synonyms: L-NNA, H-Arg(NO2)-OH

H62061-06 H62061-14
AAH62061-06EA 58.1 USD
AAH62061-06 AAH62061-14
Nω-Nitro-L-arginine 98%
Nomega-Nitro-L-arginine

N^w-Nitro-L-arginine, 98%


Purity: 98% CAS No.: 2149-70-4 Molecular Formula: C6H13N5O4 Formula Weight: 219.20 Melting Point: 257° Beilstein Reference: 1728914 MDL No.: MFCD00007033

Formula: C₆H₁₃N₅O₄
MW: 219.20 g/mol
Melting Pt: 257 °C
Storage Temperature: Ambient
MDL Number: MFCD00007033
CAS Number: 2149-70-4
EINECS: 218-418-9

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR