trans-4-(Boc-amino)cyclohexaneacetic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-(trans-4-((tert-Butoxycarbonyl)amino)cyclohexyl)acetic acid, Boc-1,4-trans-acha-oh
H62058-03
H62058-06
AAH62058-03EA
108
USD
AAH62058-03
AAH62058-06
AAH62058-MD
trans-4-(Boc-amino)cyclohexaneacetic acid 97%
trans-4-(Boc-amino)cyclohexaneacetic acid
Purity: 97%
CAS No.: 189153-10-4
Molecular Formula: C13H23NO4
Formula Weight: 257.32
MDL No.: MFCD03844588
Formula:
C₁₃H₂₃NO₄ Storage Temperature: Ambient |
MDL Number:
MFCD03844588 CAS Number: 189153-10-4 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...