4-Benzyl-N-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-aspartate 97%
Supplier: Thermo Scientific Chemicals
Synonyms:
Fmoc-Asp(OBzl)-OH, 4-Benzyl N-Fmoc-L-aspartate, N-Fmoc-L-aspartic Acid 4-Benzyl Ester, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-aspartic Acid 4-Benzyl Ester
H62011-06
H62011-14
AAH62011-06EA
77.8
USD
AAH62011-06
AAH62011-14
4-Benzyl-N-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-aspartate 97%
4-Benzyl-N-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-aspartate
N-Fmoc-L-aspartic acid 4-benzyl ester, 97%
Purity: 97%
CAS No.: 86060-84-6
Molecular Formula: C26H23NO6
Formula Weight: 445.46
Storage Temperature: 2°C to 8°C
Melting Point: 120-130°
Beilstein Reference: 4609615
MDL No.: MFCD00065630
Formula:
C₂₆H₂₃NO₆ MW: 445.47 g/mol Melting Pt: 120…130 °C Storage Temperature: Ambient |
MDL Number:
MFCD00065630 CAS Number: 86060-84-6 |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...