4-Fluoro-2-methoxyaniline 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H61965-03
H61965-06
AAH61965-03EA
172
USD
AAH61965-03
AAH61965-06
AAH61965-MD
4-Fluoro-2-methoxyaniline 95%
4-Fluoro-2-methoxyaniline
4-Fluoro-2-methoxyaniline, 95%
Purity: 95%
CAS No.: 450-91-9
Molecular Formula: C7H8FNO
Formula Weight: 141.14
Physical Form: Solid
UN No.: 2811
MDL No.: MFCD00077536
Formula:
C7H8FNO Storage Temperature: Ambient |
MDL Number:
MFCD00077536 CAS Number: 450-91-9 UN: 2811 ADR: "6.1",III |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...