5-Bromothiophene-2-carbohydrazide 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-Bromothiophene-2-carboxylic acid hydrazide

H61959-06 H61959-14
AAH61959-06EA 142 USD
AAH61959-06 AAH61959-14
5-Bromothiophene-2-carbohydrazide 98%
5-Bromothiophene-2-carbohydrazide

5-Bromothiophene-2-carboxylic acid hydrazide, 98%


Purity: 98% CAS No.: 98027-27-1 Molecular Formula: C5H5BrN2OS Formula Weight: 221.07 MDL No.: MFCD01310798

Formula: C₅H₅BrN₂OS
MW: 221.072 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01310798
CAS Number: 98027-27-1

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR