4-Methylthiazole-5-carbaldehyde 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-Methylthiazole-5-carboxaldehyde, 5-formyl-4-methyl thiazole, 4-Methyl-5-formylthiazole

H61613-03 H61613-06
AAH61613-03EA 43.6 USD
AAH61613-03 AAH61613-06
4-Methylthiazole-5-carbaldehyde 97%
4-Methylthiazole-5-carbaldehyde

4-Methylthiazole-5-carboxaldehyde, 97%


Purity: 97% CAS No.: 82294-70-0 Molecular Formula: C5H5NOS Formula Weight: 127.16 Melting Point: 74-78° MDL No.: MFCD07368277

Formula: C₅H₅NOS
MW: 127.161 g/mol
Melting Pt: 74…78 °C
Storage Temperature: Ambient
MDL Number: MFCD07368277
CAS Number: 82294-70-0

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR