Trimethylboroxine 50% (w/w) in THF ≥98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Trimethylboroxine, 50% w/w soln. in THF
CAS No.: 823-96-1
Molecular Formula: C3H9B3O3
Formula Weight: 125.54
Physical Form: Liquid
Sensitivity: Moisture Sensitive; Hygroscopic
Appearance: Colorless to yellow
Beilstein Reference: 1757008
UN No.: 2924
MDL No.: MFCD00013354
Formula:
C₃H₉B₃O₃ Density: 0.894 Storage Temperature: Ambient |
MDL Number:
MFCD00013354 CAS Number: 823-96-1 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...