6-Amino-3-chloro-1H-indazole 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Chloro-1H-indazol-6-amine

H61445-03 H61445-06
AAH61445-03EA 198 USD
AAH61445-03 AAH61445-06
6-Amino-3-chloro-1H-indazole 97%
6-Amino-3-chloro-1H-indazole

6-Amino-3-chloro-1H-indazole, 97%


Purity: 97% CAS No.: 21413-23-0 Molecular Formula: C7H6ClN3 Formula Weight: 167.60 Storage Temperature: 2°C to 8°C MDL No.: MFCD07781648

Formula: C₇H₆ClN₃
MW: 167.60 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07781648
CAS Number: 21413-23-0

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR