3-Thiophenecarbonyl chloride 97%

Supplier: Thermo Scientific Chemicals
Danger

H61428-06 H61428-14
AAH61428-06EA 209.08 USD
AAH61428-06 AAH61428-14
3-Thiophenecarbonyl chloride 97%
3-Thiophenecarbonyl chloride

Thiophene-3-carbonyl chloride, 97%


Purity: 97% CAS No.: 41507-35-1 Molecular Formula: C5H3ClOS Formula Weight: 146.59 Storage Temperature: 2°C to 8°C Flash Point: 51°(124°F) Melting Point: 51-54° UN No.: 2921 MDL No.: MFCD00130090

Formula: C₅H₃ClOS
MW: 146.588 g/mol
Melting Pt: 51…54 °C
Flash Pt: 51 °C (124 °F)
Storage Temperature: Ambient
MDL Number: MFCD00130090
CAS Number: 41507-35-1
EINECS: 255-420-9
UN: 2921
ADR: 8,II

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR