N-Carbobenzoxy-D-glutamic acid 95%

Supplier: Thermo Scientific Chemicals

Synonyms: N-Benzyloxycarbonyl-D-glutamic acid, N-Cbz-D-glutamic Acid, Z-D-Glu-OH, Cbz-D-Glu-OH

H61401-14 H61401-22
AAH61401-14EA 104 USD
AAH61401-14 AAH61401-22
N-Carbobenzoxy-D-glutamic acid 95%
N-Carbobenzoxy-D-glutamic acid

N-Benzyloxycarbonyl-D-glutamic acid, 95%


Purity: 95% CAS No.: 63648-73-7 Molecular Formula: C13H15NO6 Formula Weight: 281.26 Melting Point: 119-121° MDL No.: MFCD00063193

Formula: C₁₃H₁₅NO₆
MW: 281.26 g/mol
Melting Pt: 119…121 °C
Storage Temperature: Ambient
MDL Number: MFCD00063193
CAS Number: 63648-73-7

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR