(6-Bromo-3-pyridinyl)methanol 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Bromo-5-(hydroxymethyl)pyridine, 2-Bromo-5-pyridinemethanol, (6-Bromopyridin-3-yl)methanol
H61375-03
H61375-06
AAH61375-03EA
40.7
USD
AAH61375-03
AAH61375-06
(6-Bromo-3-pyridinyl)methanol 95%
(6-Bromo-3-pyridinyl)methanol
2-Bromo-5-pyridinemethanol, 95%
Purity: 95%
CAS No.: 122306-01-8
Molecular Formula: C6H6BrNO
Formula Weight: 188.02
Melting Point: 48-51°
MDL No.: MFCD04974508
Formula:
C₆H₆BrNO MW: 188.02 g/mol Melting Pt: 48…51 °C Storage Temperature: Ambient |
MDL Number:
MFCD04974508 CAS Number: 122306-01-8 |
Specification Test Results
Assay | 95% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...