2-(Diphenylphosphino)benzaldehyde 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(2-Formylphenyl)diphenylphosphine
H61306-03
H61306-06
AAH61306-03EA
71.9
USD
AAH61306-03
AAH61306-06
2-(Diphenylphosphino)benzaldehyde 97%
2-(Diphenylphosphino)benzaldehyde
2-(Diphenylphosphino)benzaldehyde, 97%
Purity: 97%
CAS No.: 50777-76-9
Molecular Formula: C19H15OP
Formula Weight: 290.30
Physical Form: Crystalline powder
Appearance: Yellow
Melting Point: 112-115°
MDL No.: MFCD00013367
Formula:
C₁₉H₁₅OP MW: 290.30 g/mol Melting Pt: 112…115 °C Storage Temperature: Ambient |
MDL Number:
MFCD00013367 CAS Number: 50777-76-9 |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...