Nicotinamide-N-oxide 98%

Supplier: Thermo Scientific Chemicals
Warning

H61292-14 H61292-22
AAH61292-14EA 210 USD
AAH61292-14 AAH61292-22
Nicotinamide-N-oxide 98%
Nicotinamide-N-oxide

Nicotinamide N-oxide, 98%


Purity: 98% CAS No.: 1986-81-8 Molecular Formula: C6H6N2O2 Formula Weight: 138.12 Melting Point: 291-293° dec MDL No.: MFCD00006202

Formula: C₆H₆N₂O₂
Melting Pt: 291…293 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00006202
CAS Number: 1986-81-8
EINECS: 217-859-4

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR