2-Trimethylsilylthiophene 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Thienyltrimethylsilane, Trimethyl(thiophen-2-yl)silane
H61170-06
H61170-14
AAH61170-06EA
73.5
USD
AAH61170-06
AAH61170-14
2-Trimethylsilylthiophene 97%
2-Trimethylsilylthiophene
2-(Trimethylsilyl)thiophene, 97%
Purity: 97%
CAS No.: 18245-28-8
Molecular Formula: C7H12SSi
Formula Weight: 156.32
Storage Temperature: 2°C to 8°C
Flash Point: 41°(106°F)
Refractive Index: 1.497
UN No.: 1993
MDL No.: MFCD00005415
Formula:
C₇H₁₂SSi MW: 156.32 g/mol Density: 0.926 Flash Pt: 41 °C(106 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005415 CAS Number: 18245-28-8 UN: 1993 ADR: "3",III |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...