Potassium-2,4-difluorophenyltrifluoroborate 95%
Supplier: Thermo Scientific Chemicals
Synonyms:
Potassium (2,4-difluorophenyl)trifluoroborate
H37326-03
H37326-06
AAH37326-03EA
24.2
USD
AAH37326-03
AAH37326-06
Potassium-2,4-difluorophenyltrifluoroborate 95%
Potassium 2,4-difluorophenyltrifluoroborate
Potassium 2,4-difluorophenyltrifluoroborate, 95%
Purity: 95%
CAS No.: 871231-41-3
Molecular Formula: C6H3BF5K
Formula Weight: 219.99
MDL No.: MFCD07784375
Formula:
C₆H₃BF₅K MW: 219.99 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07784375 CAS Number: 871231-41-3 |
Specification Test Results
Assay | 95% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...