Nε-trifluoroacetyl-L-lysine 97%
Supplier: Thermo Scientific Chemicals
H37051-06
H37051-14
AAH37051-06EA
27.29
USD
AAH37051-06
AAH37051-14
Nε-trifluoroacetyl-L-lysine 97%
Nepsilon-trifluoroacetyl-L-lysine
N^e-Trifluoroacetyl-L-lysine, 97%
Purity: 97%
CAS No.: 10009-20-8
Molecular Formula: C8H13F3N2O3
Formula Weight: 242.20
Storage Temperature: 2°C to 8°C
Melting Point: ca 253° dec.
Optical Rotation: +19°(c=1 in 1M HCl)
MDL No.: MFCD00037223
Formula:
C₈H₁₃F₃N₂O₃ Melting Pt: ca 253 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00037223 CAS Number: 10009-20-8 |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...