4-Vinylpyridine 96% stabilized low polymer
Supplier: Thermo Scientific Chemicals
|
Danger
|
Stabilization:
Stabilised with hydroquinone (HQ)
H36621-AC
H36621-AE
H36621-AP
AAH36621-ACEA
50.6
USD
AAH36621-AC
AAH36621-AE
AAH36621-AP
4-Vinylpyridine 96% stabilized low polymer
4-Vinylpyridine
4-Vinylpyridine, 96%, low polymer, stab. with 100ppm hydroquinone
Purity: 96%
CAS No.: 100-43-6
Molecular Formula: C7H7N
Formula Weight: 105.14
Storage Temperature: -30°C to -10°C
Flash Point: 51°(124°F)
Refractive Index: 1.5512
Boiling Point: 61°/12mm
UN No.: 3073
MDL No.: MFCD00006447
Formula:
C₇H₇N MW: 105.14 g/mol Boiling Pt: 61 °C/12 mm Density: 0.984 Flash Pt: 51 °C (124 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00006447 CAS Number: 100-43-6 EINECS: 202-852-0 UN: 3073 ADR: 6.1,II |
Specification Test Results
Assay | 96% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...