Lithium hydroxide monohydrate battery grade

Supplier: Thermo Scientific Chemicals
Danger

H36379-0B H36379-30
AAH36379-0BEA 439.44 USD
AAH36379-0B AAH36379-30
Lithium hydroxide monohydrate battery grade
Lithium hydroxide monohydrate

Lithium hydroxide monohydrate, battery grade


CAS: 1310-66-3 MDL: MFCD00149772

Formula: LiOH·1H₂O
MW: 41.96 g/mol
Boiling Pt: 920
Melting Pt: 462
Storage Temperature: Ambient
MDL Number: MFCD00149772
CAS Number: 1310-66-3
EINECS: 215-183-4
UN: 2680
ADR: 8 II
Merck Index: 13,05556

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR