Zirconium diboride ≥99.5% (metals basis excluding Hf)
Supplier: Thermo Scientific Chemicals
|
Warning
|
047030-18
047030-30
AA47030-18EA
157
USD
AA47030-18
AA47030-30
Zirconium diboride ≥99.5% (metals basis excluding Hf)
Zirconium diboride
Zirconium boride, 99.5% (metals basis excluding Hf), 15 Micron APS Powder.
Purity: 99.5%
CAS No.: 12045-64-6
Molecular Formula: B2Zr
Formula Weight: 112.84
Physical Form: 15 Micron APS Powder
Melting Point: 3000°
Optical Rotation: Zirconium diboride
UN No.: 3178
MDL No.: MFCD00064648
Formula:
B₂Zr MW: 112.84 g/mol Melting Pt: 3000 °C Storage Temperature: Ambient |
MDL Number:
MFCD00064648 CAS Number: 12045-64-6 EINECS: 234-963-5 UN: 3178 ADR: 4.1,III |
Specification Test Results
Assay | 99.5% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...