Aluminium oxide basic 32-63 µm APS HPLC flash grade
Supplier: Thermo Scientific Chemicals
046071-22
046071-36
AA46071-22EA
58.2
USD
AA46071-22
AA46071-36
Aluminium oxide basic 32-63 µm APS HPLC flash grade
Aluminium oxide basic
Aluminum oxide, basic, HPLC Flash Grade, 32-63 Micron APS Powder.
CAS No.: 1344-28-1
Molecular Formula: Al2O3
Formula Weight: 101.96
Physical Form: 32-63 Micron APS Powder
Refractive Index: 1.765
Appearance: White
Odor: Odorless
Melting Point: 2045°
Boiling Point: 2980°
MDL No.: MFCD00003424
Formula:
Al₂O₃ MW: 101.96 g/mol Boiling Pt: 2980 °C Melting Pt: 2045 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003424 CAS Number: 1344-28-1 EINECS: 215-691-6 Merck Index: 14,356 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...