Sodium telluride ≥99.9% -60 mesh

Supplier: Thermo Scientific Chemicals
Danger

041777-04 041777-09
AA41777-04EA 320 USD
AA41777-04 AA41777-09
Sodium telluride ≥99.9% -60 mesh
Sodium telluride

Sodium telluride, 99.9% (metals basis), -60 Mesh Powder.


Purity: 99.9% CAS No.: 12034-41-2 Molecular Formula: Na2Te Formula Weight: 173.58 Physical Form: -60 Mesh Powder Density: 2.90 Sensitivity: Moisture Sensitive Appearance: White Odor: Odorless Melting Point: 953° UN No.: 3134 MDL No.: MFCD00053400

Formula: Na₂Te
MW: 173.58
Melting Pt: 953 °C
Density: 2.90
Storage Temperature: Ambient
MDL Number: MFCD00053400
CAS Number: 12034-41-2
EINECS: 234-806-0
UN: 3134
ADR: 4.3 II

Order Now

Specification Test Results

Assay 99.9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR