Technocut® Disposable Scalpels, Sterile, MYCO Medical
Supplier: Myco Medical
6008T-10 6008T-11 6008T-12 6008T-12B 6008T-15 6008T-15C 6008T-20 6008T-22 6008T-23
10148-882BX 24.08 USD
10148-882 10148-884 10148-886 10148-888 10148-890 10148-892 10148-894 10148-896 10148-898
Technocut® Disposable Scalpels, Sterile, MYCO Medical
Scalpels and Scalpel Assemblies Scalpels
Technocut® Disposable Scalpels feature scalpel blades produced from high quality Swiss stainless steel that are attached to the handle by cold staking
The non-slip clear shield improves safety and provides for easy blade size identification. These scalpels have ergonomic, non-slip plastic handles with an integrated metric ruler for convenience.