KIMBLE® FLEX-COLUMN® Sample Diffusion Disc, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
420404-1000 420404-1500 420404-2500
KT420404-1000EA 41.19 USD
KT420404-1000 KT420404-1500 KT420404-2500
KIMBLE® FLEX-COLUMN® Sample Diffusion Disc, DWK Life Sciences
Chromatography Columns
Sample diffusion discs protect the gel bed from being disrupted during sample application whenever a flow adapter is not used.
A transparent acrylic ring is sealed to a 20 μm porosity HDPE disc which protects the top of the gel bed when the sample is manually applied. A crossbar on the top of the acrylic ring permits easy removal of the sample diffusion disc from the column.