Nickel(II) sulphate hexahydrate, extra pure, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Nickelous sulphate, Hexahydrate, Nickel sulphate hexahydrate

M-Clarity™ quality level = MQ200

1.06726.1000 1.06726.9050
EM1.06726.1000CS 3886 USD
EM1.06726.1000 EM1.06726.9050
Nickel(II) sulphate hexahydrate, extra pure, Supelco®
Nickel(II) sulphate hexahydrate
Formula: NiSO₄·6H₂O
MW: 262.85 g/mol
Melting Pt: 840
Density: 2.07 g/cm3 at 20 °C
Storage Temperature: Ambient
MDL Number: MFCD00149813
CAS Number: 10101-97-0
EINECS: 232-104-9
UN: 3077
ADR: 9 III
Merck Index: 13,06541

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR