Reli® Chiba Needles, Sterile, MYCO Medical
Supplier: Myco Medical
CHE22G801 CHE22G601 CHE22G501 CHE22G351 CHE22G101 CHE22G701 CHE20G601 CHE18G801 CHE20G801 CHE25G601 CHE25G801 CHE20G701 CHE18G351 CHE18G601 CHE20G351 CHE23G601 CHE23G801
10148-596BX 268.9 USD
10148-596 10148-598 10148-600 10148-602 10148-604 10148-606 10148-608 10148-610 10148-612 10148-614 10148-634 10148-636 10148-638 10148-640 10148-642 10148-644 10148-646
Reli® Chiba Needles, Sterile, MYCO Medical
Syringe Needles
Reli Chiba Needles are high quality needles with large, clear, color-coded hubs for enhanced tactile feel and easy gauge identification, and a depth stopper to aid in placement accuracy.