Aluminium oxide, fibers, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Aluminium(III) oxide, Alumina C, Aluminum oxide

M-Clarity™ quality level = MQ200

1.15754.0100 1.15754.1000
EM1.15754.0100CS 1011 USD
EM1.15754.0100 EM1.15754.1000
Aluminium oxide, fibers, Supelco®
Aluminium oxide
Formula: Al₂O₃
MW: 101.96 g/mol
Boiling Pt: 2980 °C (1013 hPa)
Melting Pt: 2045 °C
Density: 3.94 g/cm³ (20 °C)
MDL Number: MFCD00003424
CAS Number: 1344-28-1
EINECS: 215-691-6
REACH: 01-2119529248-35
Merck Index: 13,00355

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR