Aluminium hydroxide (hydrargillite), powder, EMPLURA®, Supelco®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

1.01091.1000 1.01091.9050
EM1.01091.1000CS 660 USD
EM1.01091.1000 EM1.01091.9050
Aluminium hydroxide (hydrargillite), powder, EMPLURA®, Supelco®
Aluminium hydroxide
Formula: Al(OH)₃
Melting Pt: 300 °C (–H₂O of crystall.)
Density: 2.42 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003420
CAS Number: 21645-51-2
EINECS: 244-492-7
Merck Index: 13,00342

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR