Ammonium persulfate (APS) ≥98.0% (by iodometry), Supelco®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

1.01200.1000 1.01200.5000 1.01200.9025
EM1.01200.1000CS 2125 USD
EM1.01200.1000 EM1.01200.5000 EM1.01200.9025
Ammonium persulfate (APS) ≥98.0% (by iodometry), Supelco®
Ammonium peroxodisulphate (APS)
Formula: (NH₄)₂S₂O₈
Melting Pt: 120 °C (dec.)
Density: 1.98 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003390
CAS Number: 7727-54-0
EINECS: 231-786-5
UN: 1444
ADR: 5.1,III
Merck Index: 13,00545
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f82a373f21d34e63dc94c0598f3f33ba6bbfdb2b&vid=2b6d196dbf07ce4dd528ac7b4f8456678accd3ba&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR