Petroleum spirit, 40 °C ~, EMPLURA®, Supelco®
Supplier: MilliporeSigma
|
Danger
|
M-Clarity™ quality level = MQ200
1.00915.1000
1.00915.5000
1.00915.6025
EM1.00915.1000CS
666
USD
EM1.00915.1000
EM1.00915.5000
EM1.00915.6025
Petroleum spirit, 40 °C ~, EMPLURA®, Supelco®
Petroleum spirit
Boiling Pt:
∼40 °C (1013 hPa) Density: 0.62 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00081849 CAS Number: 64742-49-0 EINECS: 265-151-9 UN: 1268 ADR: 3,I |
Specification Test Results
Boiling range (30-40°C) | ≥ 90 % (v/v) |
Free acid (as CH₃COOH) | ≤ 0.0005 % |
Density (d 20 °C/ 4 °C) | 0.623 - 0.627 |
Readily carbonizable substances | conforms |
Evaporation residue | ≤ 0.0020 % |
Water | ≤ 0.010 % |
Aromatics (as benzene) | ≤ 0.01 % |
Sulfur compounds (as S) | ≤ 0.01 % |
Al (Aluminium) | ≤ 0.00002 % |
Ba (Barium) | ≤ 0.00001 % |
Ca (Calcium) | ≤ 0.0001 % |
Cd (Cadmium) | ≤ 0.00001 % |
Co (Cobalt) | ≤ 0.00001 % |
Cr (Chromium) | ≤ 0.000003 % |
Cu (Copper) | ≤ 0.000003 % |
Fe (Iron) | ≤ 0.000003 % |
K (Potassium) | ≤ 0.00003 % |
Mg (Magnesium) | ≤ 0.00001 % |
Mn (Manganese) | ≤ 0.000003 % |
Na (Sodium) | ≤ 0.0001 % |
Ni (Nickel) | ≤ 0.000003 % |
Pb (Lead) | ≤ 0.00001 % |
Sr (Strontium) | ≤ 0.000003 % |
Zn (Zinc) | ≤ 0.00003 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...