Sand (sea sand) for analysis, purified by acid, calcinated, Supelco®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

1.07712.1000 1.07712.5000 1.07712.9025
EM1.07712.1000CS 1881 USD
EM1.07712.1000 EM1.07712.5000 EM1.07712.9010 EM1.07712.9025
Sand (sea sand) for analysis, purified by acid, calcinated, Supelco®
Sand (sea sand)
Formula: SiO₂
MW: 60.08 g/mol
Boiling Pt: >2200 °C (1013 hPa)
Melting Pt: 1713 °C
Density: 2.66 g/cm³ (20 °C)
Storage Temperature: Ambient
CAS Number: 7631-86-9
EINECS: 231-545-4

Order Now

Specification Test Results

Substances soluble in hydrochloric acid ≤ 0.2 %
Chloride (Cl) ≤ 0.005 %
Acid-soluble Iron (Fe) ≤ 0.01 %
Loss on ignition (900 °C) ≤ 0.05 %
Particle size (0.1 - 0.315 mm) ≥ 90 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR