Isoamyl alcohol (mixture of isomers), EMPLURA®, Supelco®
Supplier: MilliporeSigma
|
Warning
|
M-Clarity™ quality level = MQ200
8.22255.1000
8.22255.2500
EM8.22255.1000CS
740
USD
EM8.22255.1000
EM8.22255.2500
Isoamyl alcohol (mixture of isomers), EMPLURA®, Supelco®
Isoamyl alcohol (mixture of isomers)
Formula:
C₅H₁₂O Boiling Pt: 131 °C (1013 hPa) Melting Pt: –117 °C Density: 0.8155 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002977 CAS Number: 30899-19-5 EINECS: 250-378-8 UN: 1105 ADR: 3,III |
Specification Test Results
Assay isomers (GC, area%) | ≥ 99.0 % (a/a) |
Density (d 20 °C/ 4 °C) | 0.808 - 0.812 |
Water (K. F.) | ≤ 0.30 % |
Identity (IR) | passes test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...