tert-Butanol, EMPLURA®, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Trimethylcarbinol, tert-Butyl alcohol, 2-Methyl-2-propanol

8.22264.1000 8.22264.9025
EM8.22264.1000CS 680 USD
EM8.22264.1000 EM8.22264.5000 EM8.22264.9025
tert-Butanol, EMPLURA®, Supelco®
tert-Butanol
Formula: (CH₃)₃COH
MW: 74.12 g/mol
Boiling Pt: 82.2
Melting Pt: 25.3
Density: 0.7887 g/cm³ (20 °C)
Flash Pt: 11 °C
Storage Temperature: Ambient
MDL Number: MFCD00004464
CAS Number: 75-65-0
EINECS: 200-889-7
UN: 1120
ADR: 3 II
Merck Index: 12,01577
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=63a1b0c51eec643e5d5b7152a3e410db93d1626e&vid=999328a3c31895b190d09313b24501682921af01&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR