N,N-Dimethylformamide, EMPLURA®, Supelco®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
NSC 5356
, Formic acid dimethylamide
, Dimethylformamide
, DMFA
, DMF
, Formyldimethylamine
, N-Formyldimethylamine
M-Clarity™ quality level = MQ200
8.22275.1000
8.22275.6025
8.22275.1000
8.22275.2500
8.22275.6025
8.22275.2500
EM8.22275.1000CS
808
USD
EM8.22275.1000
CA8.22275.6025
EM8.22275.2500
EM8.22275.6025
CA8.22275.2500
CA8.22275.1000
N,N-Dimethylformamide, EMPLURA®, Supelco®
N,N-Dimethylformamide
Formula:
HCON(CH₃)₂ MW: 73.09 g/mol Boiling Pt: 153 °C (1013 hPa) Melting Pt: –61 °C Density: 0.949 g/cm³ (20 °C) Flash Pt: 57.5 °C (closed cup) Storage Temperature: Ambient |
MDL Number:
MFCD00003284 CAS Number: 68-12-2 EINECS: 200-679-5 UN: 2265 ADR: 3,III REACH: 01-2119475605-32 Merck Index: 13,03269 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...