di-Ammonium oxalate monohydrate ≥99.0% (by manganometry), Supelco®
Supplier: MilliporeSigma
|
Warning
|
Synonyms:
Ammonium oxalate monohydrate, Ammonium oxalate-1-hydrate, Oxalic acid diammonium salt monohydrate, Oxalic acid ammonium salt monohydrate, Ammonium ethanedioate monohydrate, Ammonium Oxalate, Monohydrate
M-Clarity™ quality level = MQ200
1.01190.1000
1.01190.9050
EM1.01190.1000CS
2548
USD
EM1.01190.1000
EM1.01190.9050
di-Ammonium oxalate monohydrate ≥99.0% (by manganometry), Supelco®
di-Ammonium oxalate monohydrate
Formula:
(NH₄)₂C₂O₄·H₂O Boiling Pt: 70 °C (1013 hPa) Melting Pt: >123 °C (dec.) Density: ~ 1.5 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149694 CAS Number: 6009-70-7 EINECS: 214-202-3 UN: 3288 ADR: 6.1,III Merck Index: 13,00541 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...