Copper(II) acetate monohydrate ≥99.0% (by iodometry), crystals, EMPLURA®, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Copper diacetate monohydrate, Cupric acetate monohydrate, Acetic acid copper (II) salt monohydrate, Copper(II) acetate hydrate

M-Clarity™ quality level = MQ200

1.02710.0500 1.02710.9050
EM1.02710.0500CS 3477 USD
EM1.02710.0500 EM1.02710.9050
Copper(II) acetate monohydrate ≥99.0% (by iodometry), crystals, EMPLURA®, Supelco®
Copper(II) acetate monohydrate
Formula: (H₃CCOO)₂Cu·H₂O
MW: 199.65 g/mol
Melting Pt: 115 °C
Density: 1.88 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149570
CAS Number: 6046-93-1
EINECS: 205-553-3
UN: 1759
ADR: 8,II
Merck Index: 13,02651

Order Now

Specification Test Results

Assay (iodometric) ≥ 99.0 %
In water insoluble matter ≤ 0.1 %
pH-value (2 %; water) 5.2 - 5.5
Chloride (Cl) ≤ 0.01 %
Sulfate (SO₄) ≤ 0.2 %
Fe (Iron) ≤ 0.005 %
Zn (Zinc) ≤ 0.01 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR