Petroleum spirit, 40…80 °C, EMPLURA®, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Gasoline, Petroleum ether, Petrol, Benzine

M-Clarity™ quality level = MQ200

1.01773.1000 1.01773.6025
EM1.01773.1000CS 640 USD
EM1.01773.1000 EM1.01773.5000 EM1.01773.6025
Petroleum spirit, 40…80 °C, EMPLURA®, Supelco®
Petroleum spirit
Boiling Pt: 40…80 °C (1013 hPa)
Density: 0.665 g/cm³ (15 °C)
Flash Pt: <–30 °C
Storage Temperature: Ambient
MDL Number: MFCD00081849
CAS Number: 64742-49-0
EINECS: 265-151-9
UN: 1268
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR