Absorbent, Chemizorb® Powder, EMD Millipore

Supplier: MilliporeSigma
1.02051.0500 1.02051.9025
EM1.02051.0500CS 1711 USD
EM1.02051.0500 EM1.02051.9025
Absorbent, Chemizorb® Powder, EMD Millipore
Spill Control Materials
For absorption of acids, alkalis or solvents. Thanks to its large surface area, the powder absorbs a very large amount of liquid. Low viscosity liquids are absorbed instantaneously, highly viscous media (e.g. glycerine, oils) within a few minutes. 1 kilogram absorbs: 2,5 to 4,5 litres of aqueous solutions, acids, alkalis; 3 to 4 litres of organic solvent, benzene etc. or approximately 7 litres of viscous oils.

  • Grain size <0,1 mm (85%)
  • Bulk density approx. 120 kg/m³

CAS: 61790-53-2

MDL: MFCD00132803

Merck products are not available from Avantor in all countries. Please check with your local distributor.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR