Tetrahydrofuran ≥99% (by GC), EMPLURA®, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Butylene oxide, THF, Diethylene oxide

M-Clarity™ quality level = MQ200

1.08114.1000 1.08114.2500 1.08114.6025 1.08114.6190 1.08114.9190
EM1.08114.1000CS 675 USD
EM1.08114.1000 EM1.08114.2500 EM1.08114.6025 EM1.08114.6190 EM1.08114.9190
Tetrahydrofuran ≥99% (by GC), EMPLURA®, Supelco®
Tetrahydrofuran

For full details of specification and hazards please see www.vwr.com or the VWR Chemicals catalogues

Formula: C₄H₈O
MW: 72.11 g/mol
Boiling Pt: 66 (lit.)
Melting Pt: -108.5 (lit.)
Density: 0.889 g/ml (25°C) (lit.)
Flash Pt: –21.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00005356
CAS Number: 109-99-9
EINECS: 203-726-8
UN: 2056
ADR: 3 II
REACH: 05-2114615188-48
Merck Index: 12,09351
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b2d7611caf0127fbef941994edd1d7e69487d54a&vid=2a64e21bd2e13636c3038adf839b3e9ff18509c5&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR