Calcium chloride, anhydrous, granules Reag. Ph. Eur., Supelco®

Supplier: MilliporeSigma
Warning

Synonyms: Chloro calcium, Calcium dichloride, CaCl2

M-Clarity™ quality level = MQ200

1.02378.0500 1.02378.2500
EM1.02378.0500CS 352 USD
EM1.02378.0500 EM1.02378.2500
Calcium chloride, anhydrous, granules Reag. Ph. Eur., Supelco®
Calcium chloride
Formula: CaCl₂
MW: 110.98 g/mol
Boiling Pt: 1600
Melting Pt: 772
Density: 2.174 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010903
CAS Number: 10043-52-4
EINECS: 233-140-8
REACH: 01-2119494219-28
Merck Index: 14,01659

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR