Potassium permanganate 99.0-100.5% (by manganometry), crystalline, EMPLURA®, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Permanganic acid potassium salt

M-Clarity™ quality level = MQ200

1.05080.1000 1.05080.9050
EM1.05080.1000CS 1143 USD
EM1.05080.1000 EM1.05080.9050
Potassium permanganate 99.0-100.5% (by manganometry), crystalline, EMPLURA®, Supelco®
Potassium permanganate
Formula: KMnO₄
MW: 158.03 g/mol
Melting Pt: >240 °C (dec.)
Density: 2.7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011364
CAS Number: 7722-64-7
EINECS: 231-760-3
UN: 1490
ADR: 5.1,II
Merck Index: 12,07824

Order Now

Specification Test Results

Assay (manganometric) 99.0 - 100.5 %
Identity passes test
Appearance of solution passes test
Water insoluble matter ≤ 0.2 %
Chloride (Cl) ≤ 0.01 %
Sulfate (SO₄) ≤ 0.03 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR