Ammonium benzoate ≥98.0% (by perchloric acid titration), Supelco®

Supplier: MilliporeSigma
Warning

Synonyms: Benzoic acid ammonium salt

M-Clarity™ quality level = MQ200

1.01118.1000 1.01118.9050
EM1.01118.1000CS 2434 USD
EM1.01118.1000 EM1.01118.9050
Ammonium benzoate ≥98.0% (by perchloric acid titration), Supelco®
Ammonium benzoate
Formula: C₇H₉NO₂
MW: 139.15 g/mol
Melting Pt: 198 °C (dec.)
Density: 1.26 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00013069
CAS Number: 1863-63-4
EINECS: 217-468-9
Merck Index: 12,00521

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR