Benzoic acid methyl ester, EMPLURA®, Supelco®

Supplier: MilliporeSigma
Warning

Synonyms: Methyl benzoate

M-Clarity™ quality level = MQ200

1.06059.1000 1.06059.6025
EM1.06059.1000CS 867 USD
EM1.06059.1000 EM1.06059.2500 EM1.06059.6025
Benzoic acid methyl ester, EMPLURA®, Supelco®
Benzoic acid methyl ester
Formula: C₆H₅COOCH₃
Boiling Pt: 199.6 °C (1013 hPa)
Melting Pt: –12 °C
Density: 1.0888 g/cm³ (20 °C)
Flash Pt: 83 °C
Storage Temperature: Ambient
MDL Number: MFCD00008421
CAS Number: 93-58-3
EINECS: 202-259-7
Merck Index: 13,06052
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=287e16081e44f5525ff8b4d34f0a6fece44a3073&vid=2be9af3b90e55af477c374c091cd0888dcf60b96&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR