1-Octanol, Supelco®
Supplier: MilliporeSigma
|
Warning
|
Synonyms:
n-Octanol
M-Clarity™ quality level = MQ200
1.00991.1000
1.00991.6025
EM1.00991.1000CS
794
USD
EM1.00991.1000
EM1.00991.6025
1-Octanol, Supelco®
1-Octanol
Formula:
H₃C(CH₂)₆CH₂OH MW: 130.23 g/mol Boiling Pt: 195 °C (1013 hPa) Melting Pt: –16 °C Density: 0.8246 g/cm³ (20 °C) Flash Pt: 81 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002988 CAS Number: 111-87-5 EINECS: 203-917-6 Merck Index: 13,06784 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...