Methyl ethyl ketone ≥99% (by GC), EMPLURA®, Supelco®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

1.06014.1000 1.06014.2500 1.06014.6025 1.06014.9011 1.06014.9190
EM1.06014.1000CS 531.48 USD
EM1.06014.1000 EM1.06014.2500 EM1.06014.6025 EM1.06014.9011 EM1.06014.9190
Methyl ethyl ketone ≥99% (by GC), EMPLURA®, Supelco®
Methyl ethyl ketone
Formula: C₂H₅COCH₃
Boiling Pt: 79.6 °C (1013 hPa)
Melting Pt: -86 °C
Density: 0.8054 g/cm³ (20 °C)
Flash Pt: -4 °C
Storage Temperature: Ambient
MDL Number: MFCD00011648
CAS Number: 78-93-3
EINECS: 201-159-0
UN: 1193
ADR: 3,II
REACH: 01-2119457290-43
Merck Index: 13,06097
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8b6656888f6a9a4c0c5ed3053bb2a3dcd8b3af66&vid=17cb98abf677b90f8bac752664d5c0cfd6765591&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR