Calcium nitrate tetrahydrate, Supelco®
Supplier: MilliporeSigma
|
Danger
|
M-Clarity™ quality level = MQ200
1.02120.5000
1.02120.9050
EM1.02120.5000CS
1821
USD
EM1.02120.5000
EM1.02120.9050
Calcium nitrate tetrahydrate, Supelco®
Calcium nitrate tetrahydrate
Formula:
Ca(NO₃)₂·4H₂O Melting Pt: 45 °C Density: 1.86 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149604 CAS Number: 13477-34-4 EINECS: 233-332-1 UN: 1454 ADR: 5.1,III Merck Index: 13,01687 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...