Barium chloride dihydrate, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Barium dichloride

M-Clarity™ quality level = MQ200

1.01717.1000 1.01717.9050
EM1.01717.1000CS 909 USD
EM1.01717.1000 EM1.01717.9050
Barium chloride dihydrate, Supelco®
Barium chloride dihydrate
Formula: BaCl₂·2H₂O
MW: 244.27 g/mol
Boiling Pt: 1560 °C (1013 hPa)
Melting Pt: 960 °C
Density: 3.86 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149154
CAS Number: 10326-27-9
EINECS: 233-788-1
UN: 1564
ADR: 6.1,III
Merck Index: 14,00971

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR