Sodium tungstate dihydrate ≥98% (by thermogravimetry), Extra Pure, Supelco®
Supplier: MilliporeSigma
|
Warning
|
Synonyms:
Tungstic acid sodium salt dihydrate, Sodium wolframate dihydrate, Sodium tungsten oxide dihydrate
M-Clarity™ quality level = MQ200
1.06672.1000
1.06672.9025
EM1.06672.1000CS
6641
USD
EM1.06672.1000
EM1.06672.9025
Sodium tungstate dihydrate ≥98% (by thermogravimetry), Extra Pure, Supelco®
Sodium tungstate dihydrate
Formula:
Na₂WO₄·2H₂O MW: 329.85 g/mol Melting Pt: 100 °C (–H₂O of crystall.) Density: 3.23 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149190 CAS Number: 10213-10-2 EINECS: 236-743-4 Merck Index: 12,08847 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...